Order Entry
Poland
ContactUsLinkComponent
Anti-p-HSP27 Rabbit Polyclonal Antibody [clone: S15]
Anti-p-HSP27 Rabbit Polyclonal Antibody [clone: S15]
  BWRLAP0245
Anti-p-HSP27 Rabbit Polyclonal Antibody [clone: S15]
  BWRLAP0245

 

  • Antibody type:
    Primary
  • Antigen symbol:
    p-HSP27
  • Clonality:
    Polyclonal
  • Clone:
    S15
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic phosphopeptide derived from human HSP27 around the phosphorylation site of Serine 15.

p-HSP27 (S15) pAb detects endogenous levels of HSP27 protein only when phosphorylated at Ser15

Type: Primary
Antigen: p-HSP27
Clonality: Polyclonal
Clone: S15
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Rat;(Mouse)