Anti-HSP40 Rabbit Polyclonal Antibody [clone: P303]
Catalog # BWRLBS1178
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:HSP40
- Clonality:Polyclonal
- Clone:P303
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 270-320 of Human HSP40.
HSP40 (P303)pAb detects endogenous levels of HSP40 protein.
Type: Primary
Antigen: HSP40
Clonality: Polyclonal
Clone: P303
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat