Anti-PR Rabbit Polyclonal Antibody [clone: V184]
Catalog # BWRLBS1299
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PR
- Clonality:Polyclonal
- Clone:V184
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 151-200 of Human PR.
Progesterone Receptor (V184) pAb detects endogenous levels of Progesterone Receptor protein.
Type: Primary
Antigen: PR
Clonality: Polyclonal
Clone: V184
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human