Order Entry
Poland
ContactUsLinkComponent
Multi-parameter meter (pH/mV/conductivity), handheld, Orion Star™ A325
Multi-parameter meter (pH/mV/conductivity), handheld, Orion Star™ A325
Supplier:  Thermo Orion
Certificates
undefined
Multi-parameter meter (pH/mV/conductivity), handheld, Orion Star™ A325
Supplier:  Thermo Orion

Specifications

  • Catalog No:
  • 665-0278
  • 665-0277
  • Model:
  • A325
  • A325
  • pH range:
  • –2,000...+20,000
  • –2,000...+20,000
  • pH resolution:
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • pH accuracy:
  • ±0,002
  • ±0,002
  • pH calibration:
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • mV range:
  • ±2000,0
  • ±2000,0
  • mV resolution:
  • 0,1
  • 0,1
  • mV accuracy:
  • ±0,2 mV or ±0,05% of reading, whichever is greater
  • ±0,2 mV or ±0,05% of reading, whichever is greater
  • Conductivity range:
  • 0,001 µS/cm - 3000 mS/cm
  • 0,001 µS/cm - 3000 mS/cm
  • Conductivity resolution:
  • 0,001 μS minimum, auto ranging, up to 4 significant digits
  • 0,001 μS minimum, auto ranging, up to 4 significant digits
  • Conductivity accuracy:
  • ±0,5% of reading ±1 digit
  • ±0,5% of reading ±1 digit
  • Conductivity cell constant:
  • 0.001 - 199.9cm⁻¹
  • 0.001 - 199.9 cm⁻¹
  • Conductivity calibration:
  • Up to 5 points
  • Up to 5 points
  • Resistivity range:
  • 2 Ω/cm - 100 MΩ/cm
  • 2 Ω/cm - 100 MΩ/cm
  • Resistivity accuracy:
  • 0,5% of reading ±1 digit
  • 0,5% of reading ±1 digit
  • Salinity range:
  • 0,06 - 80,00 psu / 0,05 - 42.00 ppt
  • 0,06 - 80,00 psu / 0,05 - 42.00 ppt
  • Salinity resolution:
  • 0,01 psu or 0,01 ppt minimum, auto ranging
  • 0,01 psu or 0,01 ppt minimum, auto ranging
  • Salinity accuracy:
  • 0,5% of reading ±1 digit
  • 0,5% of reading ±1 digit
  • TDS range:
  • 0,001 - 200,0 ppm
  • 0,001 - 200,0 ppm
  • TDS resolution:
  • 0,001 ppm minimum, auto ranging, up to 4 significant digits
  • 0,001 ppm minimum, auto ranging, up to 4 significant digits
  • TDS accuracy:
  • 0,5% of reading ±1 digit
  • 0,5% of reading ±1 digit
  • TDS conversion factor:
  • Linear (0,02 - 9,99)
  • Linear (0,02 - 9,99)
  • Temperature range:
  • –5,0...+105,0 °C
  • –5,0...+105,0 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,1 °C
  • ±0,1 °C
  • Temperature compensation:
  • Automatic or manual
  • Automatic or manual
  • Display:
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Battery type:
  • 4 AA (included)
  • 4 AA (included)
  • Battery life:
  • 800 h
  • 800 h
  • Power supply:
  • AC adapter or 4 AA batteries
  • AC adapter or 4 AA batteries
  • Compliance:
  • IP 67
  • IP 67
  • Data logging - log function:
  • Manual / Automatic with AUTO-READ™
  • Manual / Automatic with AUTO-READ™
  • Data logging capacity:
  • 5000 with date and time stamp
  • 5000 with date and time stamp
  • Languages:
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • IP rating:
  • IP 67
  • IP 67
  • Connections:
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe)
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe)
  • Outputs:
  • RS232, USB
  • RS232, USB
  • Input:
  • Two measuring channels
  • Two measuring channels
  • Alarm:
  • High/low limit alarm, calibration due alarm
  • High/low limit alarm, calibration due alarm
  • No. of channels:
  • 2
  • 2
  • Calibration:
  • pH with calibration editing option, relative mV (RmV), ORP, conductivity with calibration editing op
  • pH with calibration editing option, relative mV (RmV), ORP, conductivity with calibration editing op
  • Calibration points:
  • 1 point relative mV (RmV) mode, 1 point ORP mode
  • 1 point relative mV (RmV) mode, 1 point ORP mode
  • W×D×H:
  • 240×100×65 mm
  • 240×100×65 mm
  • Weight:
  • 0,5 kg
  • 0,5 kg

Specifications

Frequently Bought Together