Order Entry
Poland
ContactUsLinkComponent
Aluminium oxide MP Alumina 32-63 µm active
Aluminium oxide MP Alumina 32-63 µm active
Catalog # ICNA0202040
Supplier:  MP Biomedicals
CAS Number:  
Aluminium oxide MP Alumina 32-63 µm active
Catalog # ICNA0202040
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :101.96 g/mol
Storage Temperature:Ambient
CAS Number:1344-28-1
 :11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424