Order Entry
Poland
ContactUsLinkComponent
Anti-IGHG1 Monoclonal Antibody [clone: IG1707R]
Anti-IGHG1 Monoclonal Antibody [clone: IG1707R]
Catalog # ABNORAB00335
Supplier:  Abnova
Anti-IGHG1 Monoclonal Antibody [clone: IG1707R]
Catalog # ABNORAB00335
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    IgG
  • Antigen symbol:
    IGHG1
  • Clonality:
    Monoclonal
  • Clone:
    IG1707R
  • Conjugation:
    Unconjugated
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    3500
  • Storage buffer:
    In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
  • Storage temperature:
    Store at 4° C.
  • Immunogen:
    Original antibody is raised against native purified human IgG (Ig Gamma Chain).
  • Purification:
    Protein A purification
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal antibody raised against human IgG.

Type: Primary
Antigen: IGHG1
Clonality: Monoclonal
Clone: IG1707R
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG, kappa
Reactivity: Human