Insect cell media, powdered, IPL41
USBII8450-50
:
- Media Format:Powder
- Pk:1
- Description:IPL41 Insect medium, with L-glutamine and without calcium (powder), 50 l
- Seq:02
IPL-41 (USDA Insect Pathology Laboratory) for the large scale propagation of cells from the armyworm, Spodoptera frugiperda. IPL-41 is primarily used for the growth and maintenance of cell lines derived from lepidopterans and the propagation of viruses in these cell lines.