Order Entry
Poland
ContactUsLinkComponent
 
Anti-TUBB3 Rabbit Polyclonal Antibody
  BSBTA01857-1
undefined
Anti-TUBB3 Rabbit Polyclonal Antibody
  BSBTA01857-1

 

  • Antibody type:
    Primary
  • Antigen name:
    Tubulin Beta 3
  • Antigen symbol:
    TUBB3
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Tubulin beta-3 chain(TUBB3) detection. Tested with WB, IHC-P in Human, Mouse, Rat.

Type: Primary
Antigen: TUBB3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat