Cyfrowe refraktometry kieszonkowe do składników farmaceutycznych, seria PAL
: Atago
The PAL series for pharmaceutical ingredients (reagents) are pocket instruments delivering rapid results.
: IP 65 allows rinsing off the sample with running water.
Frequently Bought Together