Order Entry
Poland
ContactUsLinkComponent
Dichloromethane stabilised, HiPerSolv CHROMANORM® for preparative HPLC
Health
Irritant
Dichloromethane stabilised, HiPerSolv CHROMANORM® for preparative HPLC
Catalog # 84534.550
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Irritant
Dichloromethane stabilised, HiPerSolv CHROMANORM® for preparative HPLC
Catalog # 84534.550
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Metal drum
  • Pk:
    189 L
  • Seq:
    5507

Specifications

About this item

 :
 :84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.33 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
ADR:6.1,III
 :Stabilised with 2-methyl-2-butene (iso-amylene) 20 ppm
REACH:01-2119480404-41
 :GHS08
 :GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881