Order Entry
Poland
ContactUsLinkComponent
Anti-CD6 Mouse Monoclonal Antibody [clone: C6/372 3F7B5]
Anti-CD6 Mouse Monoclonal Antibody [clone: C6/372 3F7B5]
Catalog # BNUM1228-50
Supplier:  Biotium
Anti-CD6 Mouse Monoclonal Antibody [clone: C6/372 3F7B5]
Catalog # BNUM1228-50
Supplier:  Biotium

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD6
  • Clonality:
    Monoclonal
  • Clone:
    Clone C6/372 3F7B5
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgGs
  • Reactivity:
    Human
  • Gene ID:
    744366
  • Antigen synonyms:
    TP120|T12
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

CD6 is a type I transmembrane glycoprotein that contains a 24-amino acid signal sequence, three extracellular scavenger receptor cysteine-rich (SRCR) domains, a membrane-spanning domain and a 44-amino acid cytoplasmic domain. The CD6 glycoprotein is tyrosine phosphorylated during TCR-mediated T cell activation. CD6 shows significant homology to CD5. CD6 is present on mature thymocytes, peripheral T cells and a subset of B cells. Antibodies to CD6 are used to deplete T cells from bone marrow transplants to prevent graft versus host disease.

  • Purified
  • BSA-free

Type: Primary
Antigen: CD6
Clonality: Monoclonal
Clone: C6/372 3F7B5
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human