Order Entry
Poland
ContactUsLinkComponent
Anti-CD6 Mouse Monoclonal Antibody (CF568) [clone: C6/372 3F7B5]
Anti-CD6 Mouse Monoclonal Antibody (CF568) [clone: C6/372 3F7B5]
Supplier:  Biotium
Certificates
Anti-CD6 Mouse Monoclonal Antibody (CF568) [clone: C6/372 3F7B5]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC681228-100
  • BNC681228-500
  • Antigen symbol:
  • CD6
  • CD6
  • Antigen synonyms:
  • TP120|T12
  • TP120|T12
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone C6/372 3F7B5
  • Clone C6/372 3F7B5
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 744366
  • 744366
  • Isotype:
  • IgGs
  • IgGs
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together