Order Entry
Poland
ContactUsLinkComponent
Anti-BECN1 Rabbit Polyclonal Antibody
Anti-BECN1 Rabbit Polyclonal Antibody
Catalog # ABGEAP14089A
Supplier:  Abgent
Anti-BECN1 Rabbit Polyclonal Antibody
Catalog # ABGEAP14089A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Beclin 1
  • Antigen symbol:
    BECN1
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Antigen synonyms:
    Coiled-coil myosin-like BCL2-interacting protein|VPS30|ATG6|GT197|Beclin-1|beclin1
  • Size:
    100 µg
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: BECN1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human