About this item
S100 belongs to the family of calcium binding proteins. S100A and S100B proteins are two members of the S100 family. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S-100 protein has been found in normal melanocytes, Langerhans cells, histiocytes, chondrocytes, lipocytes, skeletal and cardiac muscle, Schwann cells, epithelial and myoepithelial cells of the breast, salivary and sweat glands, as well as in glial cells. Neoplasms derived from these cells also express S-100 protein, albeit non-uniformly. A large number of well-differentiated tumors of the salivary gland, adipose and cartilaginous tissue, and Schwann cell-derived tumors express S-100 protein. Almost all malignant melanomas and cases of histiocytosis X are positive for S-100 protein. Despite the fact that S-100 protein is an ubiquitous substance, its demonstration is of great value in the identification of several neoplasms, particularly melanomas and their metastases.
Type: Primary
Antigen: S100A1
Clonality: Monoclonal
Clone: 4C4.9 S100A1/1012
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Cow, Human, Mouse, Rat
Specifications
- Catalog No:
- BNC401204-100
- BNC401204-500
- Antigen symbol:
- S100A1
- S100A1
- Antigen name:
- S100
- S100
- Antigen synonyms:
- S100 Calcium Binding Protein A1|Bpb|S100 Alpha Chain|S100 Calcium Binding Protein B|NEF|S100-A1|S100 Beta Chain|S100 Calcium Binding Protein Beta Neural
- S100 Calcium Binding Protein A1|Bpb|S100 Alpha Chain|S100 Calcium Binding Protein B|NEF|S100-A1|S100 Beta Chain|S100 Calcium Binding Protein Beta Neural
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- Clone 4C4.9 S100A1/1012
- Clone 4C4.9 S100A1/1012
- Reactivity:
- Human, Bovine, Rat, Mouse
- Human, Bovine, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 515715
- 515715
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide