Anti-TAGLN2 Rabbit Polyclonal Antibody
Catalog # ABGEAP14063B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Transgelin 2
- Antigen symbol:TAGLN2
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TAGLN2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human