Anti-NEUROG2 Rabbit Polyclonal Antibody
ABGEAP14019C
: Abgent
- Antibody type:Primary
- Antigen name:Neurogenin 2
- Antigen symbol:NEUROG2
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:ngn-2|Atoh4|NGN2|bHLHa8|Math4A
- Size:100 µg
- Pk:400 µl
Type: Primary
Antigen: NEUROG2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human