Anti-HEXIM1 Rabbit Polyclonal Antibody
ABGEAP13992A
: Abgent
- Antibody type:Primary
- Antigen symbol:HEXIM1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:400 µl
Type: Primary
Antigen: HEXIM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse