Order Entry
Poland
ContactUsLinkComponent
 
Anti-TFF1 Mouse Monoclonal Antibody (CF488A) [clone: TFF1/1091]
 :  Biotium
 
Anti-TFF1 Mouse Monoclonal Antibody (CF488A) [clone: TFF1/1091]
 :  Biotium

 

  • Catalog No:
  • BNC881091-500
  • BNC881091-100
  • Antigen symbol:
  • TFF1
  • TFF1
  • Antigen name:
  • pS2
  • pS2
  • Antigen synonyms:
  • Trefoil Factor 1|Gastrointestinal trefoil protein pS2|HPS2|BCEI|Gastrointestinal Trefoil Protein|HP1A|TFF1|pNR2|Breast Cancer Estrogen Inducible Protein
  • Trefoil Factor 1|Gastrointestinal trefoil protein pS2|HPS2|BCEI|Gastrointestinal Trefoil Protein|HP1A|TFF1|pNR2|Breast Cancer Estrogen Inducible Protein
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone TFF1/1091
  • Clone TFF1/1091
  • Reactivity:
  • Monkey, Human
  • Monkey, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 162807
  • 162807
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together