Order Entry
Poland
ContactUsLinkComponent
Anti-ACTA1 (Skeletal), ACTA2 (Smooth), ACTG2 (Smooth) Mouse Monoclonal Antibody (CF488A) [clone: HHF35 MSA/953]
Anti-ACTA1 (Skeletal), ACTA2 (Smooth), ACTG2 (Smooth) Mouse Monoclonal Antibody (CF488A) [clone: HHF35 MSA/953]
Supplier:  Biotium
Certificates
Anti-ACTA1 (Skeletal), ACTA2 (Smooth), ACTG2 (Smooth) Mouse Monoclonal Antibody (CF488A) [clone: HHF35 MSA/953]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC881013-100
  • BNC881013-500
  • Antigen symbol:
  • ACTA1 (Skeletal), ACTA2 (Smooth), ACTG2 (Smooth)
  • ACTA1 (Skeletal), ACTA2 (Smooth), ACTG2 (Smooth)
  • Antigen name:
  • Muscle Specific Actin
  • Muscle Specific Actin
  • Antigen synonyms:
  • Skeletal muscle alpha actin-1|ACTC1|ACTA2|alpha skeletal muscle|ACTA1|Alpha-actin-1|Actin|Alpha-2 actin|ACTA|Cardiac muscle alpha actin-1|ACTSA
  • Skeletal muscle alpha actin-1|ACTC1|ACTA2|alpha skeletal muscle|ACTA1|Alpha-actin-1|Actin|Alpha-2 actin|ACTA|Cardiac muscle alpha actin-1|ACTSA
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone HHF35 MSA/953
  • Clone HHF35 MSA/953
  • Reactivity:
  • Rabbit, Human, Rat
  • Rabbit, Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 118127, 1288 , 500483
  • 118127, 1288 , 500483
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together