Order Entry
Poland
ContactUsLinkComponent
Anti-ANPEP Mouse Monoclonal Antibody (CF405S) [clone: APN/514]
Anti-ANPEP Mouse Monoclonal Antibody (CF405S) [clone: APN/514]
Supplier:  Biotium
Certificates
Anti-ANPEP Mouse Monoclonal Antibody (CF405S) [clone: APN/514]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC040514-100
  • BNC040514-500
  • Antigen symbol:
  • ANPEP
  • ANPEP
  • Antigen name:
  • CD13
  • CD13
  • Antigen synonyms:
  • ANPEP|PEPN|Microsomal aminopeptidase|Lap1|p150|gp150|APN|Aminopeptidase N|hAPN|Aminopeptidase M|Alanyl aminopeptidase|AP-M|Myeloid plasma membrane glycoprotein CD13
  • ANPEP|PEPN|Microsomal aminopeptidase|Lap1|p150|gp150|APN|Aminopeptidase N|hAPN|Aminopeptidase M|Alanyl aminopeptidase|AP-M|Myeloid plasma membrane glycoprotein CD13
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone APN/514
  • Clone APN/514
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 1239
  • 1239
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together