Order Entry
Poland
ContactUsLinkComponent
Anti-VIM Mouse Monoclonal Antibody (CF405S) [clone: VM452]
Anti-VIM Mouse Monoclonal Antibody (CF405S) [clone: VM452]
Supplier:  Biotium
Certificates
Anti-VIM Mouse Monoclonal Antibody (CF405S) [clone: VM452]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC040452-100
  • BNC040452-500
  • Antigen symbol:
  • VIM
  • VIM
  • Antigen name:
  • Vimentin
  • Vimentin
  • Antigen synonyms:
  • VIM
  • VIM
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone VM452
  • Clone VM452
  • Reactivity:
  • Pig, Goat, Chicken, Human, Bovine, Dog, Cat
  • Pig, Goat, Chicken, Human, Bovine, Dog, Cat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 455493
  • 455493
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together