Anti-CD6 Mouse Monoclonal Antibody (CF488A) [clone: C6/372]
Supplier: Biotium
Certificates
About this item
CD6 is a type I transmembrane glycoprotein that contains a 24-amino acid signal sequence, three extracellular scavenger receptor cysteine-rich (SRCR) domains, a membrane-spanning domain and a 44-amino acid cytoplasmic domain. The CD6 glycoprotein is tyrosine phosphorylated during TCR-mediated T cell activation. CD6 shows significant homology to CD5. CD6 is present on mature thymocytes, peripheral T cells and a subset of B cells. Antibodies to CD6 are used to deplete T cells from bone marrow transplants to prevent graft versus host disease.
Type: Primary
Antigen: CD6
Clonality: Monoclonal
Clone: C6/372
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- BNC880372-100
- BNC880372-500
- Antigen symbol:
- CD6
- CD6
- Antigen synonyms:
- TP120|T12
- TP120|T12
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 488A
- CF® 488A
- Clone:
- Clone C6/372
- Clone C6/372
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 744366
- 744366
- Isotype:
- IgG1
- IgG1
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together