Order Entry
Poland
ContactUsLinkComponent
 
Anti-CD6 Mouse Monoclonal Antibody (Biotin) [clone: C6/372]
 :  Biotium
 
Anti-CD6 Mouse Monoclonal Antibody (Biotin) [clone: C6/372]
 :  Biotium

 

  • Catalog No:
  • BNCB0372-100
  • BNCB0372-500
  • Antigen symbol:
  • CD6
  • CD6
  • Antigen synonyms:
  • TP120|T12
  • TP120|T12
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone C6/372
  • Clone C6/372
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 744366
  • 744366
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together