Order Entry
Poland
ContactUsLinkComponent
 
Anti-Progesterone Mouse Monoclonal Antibody (CF640R) [clone: 6-5E-3F]
 :  Biotium
 
Anti-Progesterone Mouse Monoclonal Antibody (CF640R) [clone: 6-5E-3F]
 :  Biotium

 

  • Catalog No:
  • BNC400236-100
  • BNC400236-500
  • Antigen name:
  • Progesterone
  • Progesterone
  • Antigen synonyms:
  • Pregn 4 ene 3 20 dione
  • Pregn 4 ene 3 20 dione
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone 6-5E-3F
  • Clone 6-5E-3F
  • Reactivity:
  • Multiple
  • Multiple
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together