Order Entry
Poland
ContactUsLinkComponent
Anti-MFGE8 Mouse Monoclonal Antibody (CF405S) [clone: MFG-06]
Anti-MFGE8 Mouse Monoclonal Antibody (CF405S) [clone: MFG-06]
Supplier:  Biotium
Certificates
Anti-MFGE8 Mouse Monoclonal Antibody (CF405S) [clone: MFG-06]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC040227-500
  • BNC040227-100
  • Antigen symbol:
  • MFGE8
  • MFGE8
  • Antigen name:
  • Milk Fat Globulin
  • Milk Fat Globulin
  • Antigen synonyms:
  • Lactadherin|EDIL1|Lactadherin precursor|MFGM|Medin|MFGE8|Breast epithelial antigen BA46|Milk fat globule-EGF factor 8 (MFG-E8)|HMFG
  • Lactadherin|EDIL1|Lactadherin precursor|MFGM|Medin|MFGE8|Breast epithelial antigen BA46|Milk fat globule-EGF factor 8 (MFG-E8)|HMFG
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone MFG-06
  • Clone MFG-06
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3745
  • 3745
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together