Order Entry
Poland
ContactUsLinkComponent
Anti-THY1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: F15-42-1]
Anti-THY1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: F15-42-1]
Catalog # BNUM0194-50
Supplier:  Biotium
Anti-THY1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: F15-42-1]
Catalog # BNUM0194-50
Supplier:  Biotium

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Thy1 Cell Surface Antigen
  • Antigen symbol:
    THY1
  • Clonality:
    Monoclonal
  • Clone:
    Clone F15-42-1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Monkey,
    Human
  • Gene ID:
    644697
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

CD90 (Thy-1) is an 18-35 kDa GPI-anchored plasma membrane glycoprotein expressed in many cell types, such as in hematopoietic cells and neurons, connective tissues, various fibroblast and stromal cell lines, tumor endothelial cell lines and others. It is involved in T-cell activation, cellular adhesion, proliferation and migration, neurite outgrowth, wound healing, apoptosis, and fibrosis. CD90 participates in multiple signaling cascades and its effects are tissue- and cell type-specific. It often functions as an important regulator of cell-cell and cell-matrix interactions.

Type: Primary
Antigen: THY1
Clonality: Monoclonal
Clone: F15-42-1
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey