About this item
This MAb reacts with an N-terminal epitope (aa 16-25) of both wild type and mutated p53. Mutation and/or allelic loss of p53 is one of the causes of a variety of mesenchymal and epithelial tumors. If it occurs in the germ line, such tumors run in families. In most transformed and tumor cells the concentration of p53 is increased 51000 fold over the minute concentrations (1000 molecules cell) in normal cells, principally due to the increased half-life (4 h) compared to that of the wild-type (20 min). p53 Localizes in the nucleus, but is detectable at the plasma membrane during mitosis and when certain mutations modulate cytoplasmic/nuclear distribution. Mutations arise with an average frequency of 70% but incidence varies from zero in carcinoid lung tumors to 97% in primary melanomas. High concentrations of p53 protein are transiently expressed in human epidermis and superficial dermal fibroblasts following mild ultraviolet irradiation. Positive nuclear staining with p53 antibody has been reported to be a negative prognostic factor in breast carcinoma, lung carcinoma, colorectal, and urothelial carcinoma. Anti-p53 positivity has also been used to differentiate uterine serous carcinoma from endometrioid carcinoma as well as to detect intratubular germ cell neoplasia.
Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: BP53-12
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Chicken, Dog, Hamster, Human, Monkey
Specifications
- Catalog No:
- BNC400013-500
- BNC400013-100
- Antigen symbol:
- TP53
- TP53
- Antigen name:
- p53
- p53
- Antigen synonyms:
- Transformation Related Protein 53 (TRP53)|Tumor Protein p53|BCC7|Cellular Tumor Antigen p53|Tumor Suppressor p53|TP53|LFS1|Antigen NY-CO-13
- Transformation Related Protein 53 (TRP53)|Tumor Protein p53|BCC7|Cellular Tumor Antigen p53|Tumor Suppressor p53|TP53|LFS1|Antigen NY-CO-13
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- Clone BP53-12
- Clone BP53-12
- Reactivity:
- Hamster, Monkey, Chicken, Human, Dog
- Hamster, Monkey, Chicken, Human, Dog
- Host:
- Mouse
- Mouse
- Gene ID:
- 654481
- 654481
- Isotype:
- IgG2a
- IgG2a
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide