Order Entry
Poland
ContactUsLinkComponent
 
Anti-MLANA Mouse Monoclonal Antibody (Biotin) [clone: M2-7C10]
 :  Biotium
 
Anti-MLANA Mouse Monoclonal Antibody (Biotin) [clone: M2-7C10]
 :  Biotium

 

  • Catalog No:
  • BNCB0009-100
  • BNCB0009-500
  • Antigen symbol:
  • MLANA
  • MLANA
  • Antigen name:
  • Melan A
  • Melan A
  • Antigen synonyms:
  • MLAN-A|Antigen LB39-AA|Melanoma antigen recognized by T-cells 1|Antigen SK29-AA|MLANA
  • MLAN-A|Antigen LB39-AA|Melanoma antigen recognized by T-cells 1|Antigen SK29-AA|MLANA
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone M2-7C10
  • Clone M2-7C10
  • Reactivity:
  • Horse, Human
  • Horse, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 154069
  • 154069
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together