Order Entry
Poland
ContactUsLinkComponent
Bovine hyaluronidase (from Testes), MP Biomedicals
Bovine hyaluronidase (from Testes), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
Bovine hyaluronidase (from Testes), MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    37326-33-3
  • Source:
    Testes
  • Species:
    Bovine
  • Storage conditions:
    −20 °C
  • Form:
    Tan lyophilized powder
  • Enzyme name:
    Hyaluronidase
  • Enzyme activity:
    ≥300 USP u/mg
  • Enzyme synonyms:
    Hyaluronate Glycanohydrolase, Hyaluronate lyase, chondroitinase, Hyaluronoglucosaminidase Hyaluronate 4-glycanohydrolase
  • Molecular weight:
    55 kDa
  • pH:
    4,5 - 6,0

Specifications

Frequently Bought Together