
Potassium iodide, white crystalline powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7681-11-0
:
Formula:Kl
MDL Number:MFCD00011405
:166 g/mol
Melting Pt:681 °C (lit.)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- pH (5% aq soln):6 - 9,2
Specification Test Results
Frequently Bought Together