Ampicillin sodium salt
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:69-52-3
:
Formula:C₁₆H₁₈N₃NaO₄S
MDL Number:MFCD00064313
:371.39 g/mol
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Synonyms:D(-)-α-Aminobenzylpenicillin sodium salt
- Storage:Store at +4 °C
- Loss on Drying:2%
- pH:8 - 10 (1% aq soln)
- pka:pKa = 2,5 (-COOH) pKa = 7,3 (-NH2) at 25 °C
- Potency:800 μg/mg
Specification Test Results
Frequently Bought Together