Starch, soluble (from potato), powder, Reagent Grade
:9005-84-9
:
:(C₆H₁₀O₅)n
:MFCD00082026
:162.14 g/mol
:Ambient
- Identity Test:Passes
- Source:Potato
- FTIR:Conforms to Standard
- Moisture Content:20%
- pH (2% aq soln):5 - 7
Frequently Bought Together