Bromophenol blue sodium salt, dark green powder ACS
Catalog # ICNA0215250605
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Clarity of Solution:Passes
- Visual Transition Interval:pH 3.0 (yellow) to pH 4.6 (blue)
Specification Test Results
About this item
:
:691.95 g/mol
Storage Temperature:Ambient
CAS Number:34725-61-6
:62625-28-9
Formula:C₁₉H₉Br₄NaO₅S
MDL Number:MFCD00013793