Prostaglandin E2 ≥98%
: PGE₂
:363-24-6
:
:C₂₀H₃₂O₅
:MFCD00077861
:352.47 g/mol
- Environmentally Preferable:
- Purity:≥98%
- Synonyms:Dinoprostone; PGE2
- Formal Name:9-oxo-11α,15S-dihydroxy-prosta-5Z,13E-dien-1-oic acid
- Formulation:A Crystalline Solid
- SMILES:O=C1[C@H](C/C=C\CCCC(O)=O)[C@@H](/C=C/[C@@H](O)CCCCC)[C@H](O)C1
- InChI Code:1S/C20H32O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h4,7,12-13,15-17,19,21,23H,2-3,5-6,8-11,14H2,1H3,(H,24,25)/b7-4-,13-12+/t15-,16+,17+,19+/m0/s1
- InChI Key:XEYBRNLFEZDVAW-ARSRFYASSA-N
- Storage:−20 °C
- Shipping:Room temperature in continental US; may vary elsewhere
- Stability:2 years from the QC date provided on the Certificate of Analysis, when stored properly
Frequently Bought Together