Potassium chloride 99.0-100.5%, crystalline powder ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7447-40-7
:
Formula:KCl
MDL Number:MFCD00011360
:74.55 g/mol
Storage Temperature:Ambient
Specification Test Results
- Assay:99,0 - 100,5%
- pH:5,4 - 8,6 (5% aq soln)
- Insoluble Matter:≤0,005%
- Iodide:≤0,002%
- Bromide:≤0,01%
- Chlorate and Nitrate:≤0,003%
- Phosphate:≤5 ppm
- Sulphate:≤0,001%
- Heavy Metals (Pb):5 ppm;
- Iron:≤3 ppm
- Calcium:≤0,002%
- Magnesium :≤0,001%
- Sodium :≤0,005%
Specification Test Results
Frequently Bought Together