Anti-DPP49 Mouse Monoclonal Antibody (Biotin) [clone: TP1/19]
ABNOMAB13860
: Abnova
- Antibody type:Primary
- Antigen name:Dipeptidylpeptidase 49
- Antigen symbol:DPP49
- Clonality:Monoclonal
- Clone:TP1/19
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Gene ID:1803
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against human DPP4.
Type: Primary
Antigen: DPP49
Clonality: Monoclonal
Clone: TP1/19
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human