Order Entry
Poland
ContactUsLinkComponent
Anti-DPP49 Mouse Monoclonal Antibody (APC) [clone: TP1/19]
Anti-DPP49 Mouse Monoclonal Antibody (APC) [clone: TP1/19]
Catalog # ABNOMAB13859
Supplier:  Abnova
Anti-DPP49 Mouse Monoclonal Antibody (APC) [clone: TP1/19]
Catalog # ABNOMAB13859
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Dipeptidylpeptidase 49
  • Antigen symbol:
    DPP49
  • Clonality:
    Monoclonal
  • Clone:
    TP1/19
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Gene ID:
    1803
  • Size:
    100 reactions
  • Pk:
    100 Assays

Specifications

About this item

Mouse monoclonal antibody raised against human DPP4.

Type: Primary
Antigen: DPP49
Clonality: Monoclonal
Clone: TP1/19
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human