Anti-AFP Mouse Monoclonal Antibody [clone: C2]
ABNOMAB13264
: Abnova
- Antibody type:Primary
- Antigen name:AFP monoclonal antibody, clone C2
- Antigen symbol:AFP
- Clonality:Monoclonal
- Clone:C2
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Gene ID:174
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against native human AFP.
Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: C2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human