Order Entry
Poland
ContactUsLinkComponent
Anti-AFP Mouse Monoclonal Antibody [clone: C2]
Anti-AFP Mouse Monoclonal Antibody [clone: C2]
Catalog # ABNOMAB13263
Supplier:  Abnova
Anti-AFP Mouse Monoclonal Antibody [clone: C2]
Catalog # ABNOMAB13263
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AFP monoclonal antibody, clone C2
  • Antigen symbol:
    AFP
  • Clonality:
    Monoclonal
  • Clone:
    C2
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Gene ID:
    174
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against native human AFP.

Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: C2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human