Anti-HSPD1/780 Mouse Monoclonal Antibody [clone: HSPD1/780]
Catalog # ABNOMAB13312
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:HSPD1/780
- Clonality:Monoclonal
- Clone:HSPD1/780
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Epitope:Full-length
- Gene ID:3329
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant human HSPD1.
Type: Primary
Antigen: HSPD1/780
Clonality: Monoclonal
Clone: HSPD1/780
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human