Order Entry
Poland
ContactUsLinkComponent
Rat IgG2a Isotype Control [clone: 2A3] (APC (Allophycocyanin)-Cy7®)
Rat IgG2a Isotype Control [clone: 2A3] (APC (Allophycocyanin)-Cy7®)
Catalog # 25-4321-U100
Supplier:  Tonbo Biosciences
Rat IgG2a Isotype Control [clone: 2A3] (APC (Allophycocyanin)-Cy7®)
Catalog # 25-4321-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    2A3
  • Conjugation:
    APC (Allophycocyanin)-Cy7®
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a
  • Storage temperature:
    +2 °C to +8 °C, protected from light
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

The A3 immunoglobulin is useful as an isotype-matched control. The 2A3 immunolglobulin binds to TNP (trinitrophenol), a hapten not expressed on mouse or human cells. It is used as an isotype control for rat IgG2a antibodies.

Type: Control
Antigen: Isotype Control
Clonality: Monoclonal
Clone: 2A3
Conjugation: APC-Cyanine7
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: N/A