Order Entry
Poland
ContactUsLinkComponent
Nitrile gloves, EcoSHIELD™ Eco Nitrile PF 250
Nitrile gloves, EcoSHIELD™ Eco Nitrile PF 250
Supplier:  BIOZYM
Certificates
undefined
Nitrile gloves, EcoSHIELD™ Eco Nitrile PF 250
Supplier:  BIOZYM
Specifications
  • Catalog No:
  • BZYM743135X_U
  • 112-4687
  • BZYM743137X_U
  • BZYM743138X_U
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Finish:
  • Textured fingertip
  • Textured fingertip
  • Textured fingertip
  • Textured fingertip
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,08 mm
  • 0,08 mm
  • 0,08 mm
  • 0,08 mm
  • Finger thickness:
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • Palm thickness:
  • 0,10 mm
  • 0,10 mm
  • 0,10 mm
  • 0,10 mm
  • EN norm:
  • EN ISO 374-1 Type B (JKPT), EN ISO 374-5 Virus
  • EN ISO 374-1 Type B (JKPT), EN ISO 374-5 Virus
  • EN ISO 374-1 Type B (JKPT), EN ISO 374-5 Virus
  • EN ISO 374-1 Type B (JKPT), EN ISO 374-5 Virus