Order Entry
Poland
Orders LinkContactUsLinkComponent
Sampling cannulae
Sampling cannulae
Sampling cannulae
Sampling cannulae
Standard cannula are unique, proprietary configurations for most manual and automated sampling applications.
  • Manufactured in both electropolished 316 stainless steel or PEEK
  • Can accommodate all standard tip filters
  • Optional stoppers set the proper sampling position
The USP defines the sampling point as: 'A zone half way between the top of the media and top of the paddle or basket not less than 1 cm from the vessel wall. The sampling position is critical but so too is the type of sampling cannula (also known as a probe) used.'
Product Family Options
Agilent/VanKel/Varian compatible cannulae
  • Description
    Material
    Ø×L
  • Bent cannula with Luer lock
    Stainless steel 316
    3,2×120 mm
    1 * 1 SZT
    1 * 1 SZT
  • Bent cannula with Luer lock
    Stainless steel 316
    3,2×195 mm
    1 * 1 SZT
    1 * 1 SZT