Propargyl chloride 98%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 3-Chloropropyne, 3-Chloro-1-propyne
- CAS Number:624-65-7
- Formula:C₃H₃Cl
- MDL Number:MFCD00000980
- Molecular Weight:74.51 g/mol
- Boiling Pt:58…61 °C (760 mmHg)
- Melting Pt:–77 °C
- Density:1.03 g/cm³
- Flash Pt:19 °C
- Storage Temperature:Refrigerator
- UN:3286
- ADR:3,II
2 Options Available Below
Product Details & Documents
Synonyms: 3-Chloropropyne, 3-Chloro-1-propyne
- CAS Number:624-65-7
- Formula:C₃H₃Cl
- MDL Number:MFCD00000980
- Molecular Weight:74.51 g/mol
- Boiling Pt:58…61 °C (760 mmHg)
- Melting Pt:–77 °C
- Density:1.03 g/cm³
- Flash Pt:19 °C
- Storage Temperature:Refrigerator
- UN:3286
- ADR:3,II
Documents
ACRO45184
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Colorless to yellow to light brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4340 to 1.4370 (20°C, 589 nm)
- GC:≥97.5 %



