Order Entry
Poland
ContactUsLinkComponent
(±)-1-(1-Naphthyl)ethylamine 98%
Irritant
(±)-1-(1-Naphthyl)ethylamine 98%
Certificates
undefined
Irritant
(±)-1-(1-Naphthyl)ethylamine 98%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.6220 to 1.6240 (20°C, 589 nm)
  • Titration with HClO4:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together