Potassium fluoride ACS
:7789-23-3
:
:KF
:MFCD00011398
:58.1 g/mol
:1505 °C (1013 hPa)
:860 °C
:2,48 g/cm³ (20 °C)
:Ambient
:1812
:6.1,III
:GHS06
:GHS05
- Appearance:White Crystalline powder
- Titration after Ion exchange:≥99.0 %
- Chloride (Cl):≤0.005 %
- Sulfate (SO4):≤0.005 %
- Potassium fluosilicate:≤0.1 % (K2SiF6)
- Iron (Fe):≤0.001 %
- Sodium (Na):≤0.2 %
- Heavy metals (as Pb):≤0.001 %
- Titrable acid:≤0.03 meq/g
- Titrable base:≤0.01 meq/g
Frequently Bought Together