
Bismuth sodium trioxide ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12232-99-4
:
Formula:BiNaO₃
MDL Number:MFCD00003474
:279.97 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to brown Solid
- Titration with KMnO4:≥80.0 %
- Chloride (Cl):≤0.002 %
- Manganese (Mn):≤5 ppm
- Oxidizing efficiency:≥99.6 %
Specification Test Results
Frequently Bought Together