Order Entry
Poland
ContactUsLinkComponent
4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde 97%
Irritant
4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde 97%
Catalog # ACRO447430050
CAS Number:  
undefined
Irritant
4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde 97%
Catalog # ACRO447430050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Off-white to light brown Powder
  • Infrared spectrum:
    Conforms
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :189.02 g/mol
Melting Pt:71…76 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:287917-96-8
Formula:C₅H₅BrN₂O
MDL Number:MFCD00103235