(±)-1,2-Propanediol ACS
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane
- CAS Number:57-55-6
- Formula:CH₃CH(OH)CH₂OH
- MDL Number:MFCD00064272
- Molecular Weight:76.1 g/mol
- Boiling Pt:187 °C (760 mmHg)
- Melting Pt:–60 °C
- Density:1.03 g/cm³
- Flash Pt:99 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane
- CAS Number:57-55-6
- Formula:CH₃CH(OH)CH₂OH
- MDL Number:MFCD00064272
- Molecular Weight:76.1 g/mol
- Boiling Pt:187 °C (760 mmHg)
- Melting Pt:–60 °C
- Density:1.03 g/cm³
- Flash Pt:99 °C
- Storage Temperature:Ambient
Documents
ACRO44741
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear viscous liquid
- Water:≤0.2 %
- GC:≥99.5 %
- Color scale:≤10 APHA
- Residue after ignition:≤0.005 %
- Chloride (Cl):≤1 ppm
- Titrable acid:≤0.0005 meq/g
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



