D-(+)-Glucose-6-phosphate sodium salt 98%
Catalog # ACRO446980010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Water:≤6 %
- Specific optical rotation:+31° to +35° (20°C, 589 nm) (c=10 in H2O)
- Titration with HClO4:97.5 to 102.5 %
Specification Test Results
About this item
:
:282.12 g/mol
Melting Pt:192…197 °C
Storage Temperature:Refrigerator
CAS Number:54010-71-8
:MFCD00136039
Formula:C₆H₁₂NaO₉P
MDL Number:MFCD00006611